3M 11515-00000 2.5 Magnifier Safety Eyewear, Clear Lens
Representative image only.
3M 11515-00000 2.5 Magnifier Safety Eyewear, Clear Lens
3M | Mfr: 11515-00000-20| Item: 8647208
Safety glasses for those who need the assistance of a reading lens
$17.39CAD / Each
BUY MORE & SAVE
  • 1-9$17.39 CAD / Each
  • 10+$15.65 CAD / Each
$17.39CAD / Each
BUY MORE & SAVE
  • 1-9$17.39 CAD / Each
  • 10+$15.65 CAD / Each
Product Description
More About this Item
Large lens size offers maximum protection while allowing for reading magnification at close range. Placement of magnifying lens, like a bifocal, reduces headaches and eyestrain. Wraparound polycarbonate construction is designed to resist impacts and block 99.9% of UV rays. Flexible temples and universal-fit nosebridge.
Compliance: ANSI Z87.1-2003, CSA Z94.3
Canadian Standards Association
Specifications
  • Frame ColorBlack
  • Lens ColorClear
  • ComplianceANSI Z87.1-2003; CSA Z94.3
  • Magnification+2.5
  • Description2.5 Magnifier Safety Eyewear, Clear Lens