Value Plastics
Value Plastics
Value Plastics, a Nordson Company, provides precision plastic tubing components including luer connectors, luer accessories, tube-to-tube fittings, threaded connectors, blood pressure and bondable fittings. Their fittings and connectors are designed for a wide range of tubing applications. Value Plastics uses a variety of engineering resins to manufacture its products. All luer fittings and connectors are manufactured to interconnect to ISO standards 594-1 and 594-2. All fittings and connectors are also manufactured in a cleanroom ISO 14644-1 Level 8 environment. Many materials are available that satisfy USP Class VI criteria. Resins free from animal derivatives are also available.
APPLY FILTERS
filter byclear all
36 RESULTS
SORT BY
Showing 1 - 24 of 36
2>