Industry
Max Graduation (mL)
Capacity (oz)
Capacity (mL)
Graduation Interval (mL)
Cap / Stopper Size
Capacity (mL)
Min Graduation (mL)
TriForest
TriForest
TriForest labware is designed by a team of biologists and plastics engineers for use in life science research and biotechnology. Manufactured from premium plastics such as polycarbonate (PC), an alternative to glass ideal for cell culture and media. Products you can find in this collection are sterile bottles, lab bottles, wide-mouth bottles, scientific flasks, Erlenmeyer flasks, Erlenmeyer flask clamps, Fernbach flasks, culture tubes, serum vials, and more. Designed for microbiology and molecular biology, all TriForest labware is leakproof, optically clear, shatterproof, autoclavable, and gamma irradiated.