In Stock
Length (in)
Temperature Points (° C)
Tough-Tags
Tough-Tags
Tough-Tags peel-off labels fit microcentrifuge tube tops and other containers. Withstand steam-autoclaving, boiling water baths, freezer temperatures, organic solvents, and caustic agents without peeling off. We are a Tough-Tags authorized distributor.