Breakthrough, automated processing from bulk to test portions in 15 minutes or less
Vernon Hills, IL, May 18, 2023—The new Cole-Parmer CG-900 Cryo-Blade™ Cryogenic Grinder is an ultra-high-capacity cryogenic grinder for comminution and processing capability up to 1 kg. High-throughput capability saves valuable time for the laboratory and meets Food and Drug Administration (FDA) regulations. The Cryo-Blade automates dispensing of liquid nitrogen in 15 minutes or less, eliminating lengthy processes and the need to handle and manipulate liquid nitrogen. It is ideal for analysis of organic pollutants in food products (pesticide residues) and QuEChERS.
The Cryo-Blade automatically dispenses liquid nitrogen to allow the sample to reach cryogenic temperatures. Only 1 g is needed as a representative test sample from a 1 kg ground batch. The grinder then activates a grinding phase to pulverize samples, resulting in repeatable and reproducible sample comminution. When liquid nitrogen is not desired or available, the unit can be used with dry ice to achieve desired results.
With this unique cryogenic grinder, automatic and programmed modes are available. In automatic mode, parameters are set based on the volume of the sample. Programmed mode allows the user to save up to 10 methods. The cycle can be executed in 15 minutes when using liquid nitrogen, or 30 minutes when using dry ice. The intuitive interface includes 12 languages.
Additional features include an insulated outer bowl, mechanical locking lever, and sensors that provide the user with a safe, consistent, and reliable sample with each use. The removable 5 L inner bowl can accommodate samples up to 1 kg and allows samples to be stored in the freezer, giving the user the option of using a second bowl to prepare a new batch. The outer bowl is insulated to always allow safe touch, although using protective gloves is recommended.
View the complete line of Cole-Parmer CG-900 Cryo-Blade™ Cryogenic Grinders at coleparmer.ca