Cole-Parmer® VapLock™ Solvent Delivery Cap Kits

Reduce chemical evaporation and exposure to hazardous vapors on multiple carboys
  • Less expensive than ordering individual components
  • Color coded caps allow easy solvent identification
  • Choose from FFKM, EPDM, or Viton® valve seal options
  • Ideal for quaternary systems
Create a closed system using solvent reservoir caps to prevent hazardous vapors from escaping into the lab. Caps accept standard 1/4"-28 UNF(M) fittings to easily connect semi-rigid tubing. Anti-twist design allows the inner body of the cap to rotate independently preventing tubes from tangling when caps are removed or attached. Caps are constructed of PTFE body and PP collar* and are autoclavable. Caps with integrated air inlet valve and filter permit air to enter the bottle for vacuum displacement while solvent is pumped to the instrument. Valves are made of PTFE and 300-series SS components and feature a 50 µm PTFE filter to reduce particulate contamination.
$325.25 - $446.44CAD / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Solvent Delivery Cap Kit, for HPLC Reverse Phase
$325.25CAD / Each
COMPARE
Solvent Delivery Cap Kit, for HPLC Normal Phase
$446.44CAD / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat