Oakton® Ion 700 Benchtop Meters
Designed for today's crowded laboratory benchtops!
  • Compact footprint is over 40% smaller than other benchtop meters
  • Large display permits easier readout viewing.
Oakton benchtop 700-series meters are designed for use in crowded laboratory settings. Compact footprint of less that 6" x 7" (16 x 18 cm) conserves valuable space, while the large, dual-display LCD provides excellent visibility—even from a distance.
Ion 700 meter measures pH, mV, relative mV, ion concentration and temperature. Automatic temperature compensation (ATC) maintains reading accuracy even with fluctuating temperatures. Models feature five-point pH calibration with automatic buffer recognition for both USA (pH 1.68, 4.01, 7.00, 10.01, 12.45) and NIST (pH 1.68, 4.01, 6.86, 9.18, 12.45) pH buffers. Meters additionally feature ion concentration calibration of up to 5 points.
Meter features include Auto-Hold, °C measurement, and easy recall of electrode slope or offset. The convenient pull-out quick reference card keeps procedures handy at all times. Use 700-series meters with any pH, ORP, or ISE electrodes with a BNC connector.
$1,181.66 - $1,829.95CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Max ISE Probe Included Description
Availability Pricing
COMPARE
1999 ppm pH and ATC probes Ion 700 Benchtop Meter with Probes
$1,317.21CAD / Each
COMPARE
1999 ppm None Ion 700 Benchtop Meter with electrode stand
$1,288.48CAD / Each
COMPARE
1999 ppm pH/ATC all in one probe Ion 700 Benchtop Meter with \"all-in-one\" pH electrode
$1,181.66CAD / Each
COMPARE
1999 ppm pH and ATC probes Ion 700 Benchtop Meter with Probes and NIST Calibration
$1,829.95CAD / Each
COMPARE
1999 ppm pH/ATC all in one probe Ion 700 Benchtop Meter with Stand and NIST Calibration
$1,516.12CAD / Each
COMPARE
1999 ppm pH/ATC all in one probe Ion 700 Benchtop Meter w/ All-In-One pH Electrode and NIST Cal
$1,698.82CAD / Each