Environmental Express Oakton® pH 700 Benchtop pH Meter
Designed for today's crowded laboratory benchtops!
  • Compact footprint is over 40% smaller than other benchtop meters
  • Large display permits easier readout viewing
The easy-to-use, microprocessor-based pH 700 benchtop meter measures pH, mV, relative mV, and temperature. The large, dual-display LCD provides excellent visibility—even from a distance. Automatic temperature compensation (ATC) maintains reading accuracy even with fluctuating temperatures. Five-point pH calibration, with automatic buffer recognition for both USA (pH 1.68, 4.01, 7.00, 10.01, 12.45) and NIST (pH 1.68, 4.01, 6.86, 9.18, 12.45) pH buffers, makes for easy calibration no matter which requirement you need to meet.
Meter features HOLD, ATC, °C measurement, recall of electrode slopand offset, and electrode diagnostics. The convenient pull-out quick reference card keeps procedures handy at all times. Meter accepts any pH or ORP electrodes with a BNC connector.
$937.81 - $1,707.66CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Max pH (pH) Min pH (pH) Description
Availability Pricing
COMPARE
16 -2 Benchtop Meter with "All-in-One" pH Electrode
$1,186.23CAD / Each
COMPARE
16 -2 Benchtop Meter with "All-in-One" pH Electrode and NIST Calibration
$1,458.80CAD / Each
COMPARE
16 -2 Benchtop Meter with Probes and Stand
$1,237.06CAD / Each
COMPARE
16 -2 Benchtop Meter with Probes, Stand, and NIST Calibration
$1,707.66CAD / Each
COMPARE
16 -2 Benchtop Meter with Probe Stand
$937.81CAD / Each
COMPARE
16 -2 Benchtop Meter with Probe Stand and NIST Calibration
$1,463.22CAD / Each