Oakton® Automatic Temperature Compensation (ATC) Probes
Provides ATC for most current Oakton pH meters
  • Ensure optimal pH and ion measurement with ATC probes
Corrosion-resistant stainless steel 30 KΩ thermistor features durable polymer handle and 3.3 ft (1-m) cable. Probes measure 88-mm L x 3-mm dia; handle adds another 67 mm to the total length. Temperature range is 0 to 100°C.
$93.56 - $233.53CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
ATC probe for pH 600, pH 610, pH 620, PC 650, PD 650, PCD 650 meters, 1-m
$233.53CAD / Each
COMPARE
ATC probe for pH 600, pH 610, pH 620, PC 650, PD 650, PCD 650 meters, 3-m
$153.23CAD / Each
COMPARE
ATC probe f/ pH 5, 6, 11, 110, 510, 1100, 2100 and Ion 6 & 510
$93.56CAD / Each
COMPARE
ATC Probe for 150-, 300-, 450-series meters
$130.40CAD / Each