Cole-Parmer® Deuterium Lamps
Economical replacement deuterium lamps save you money
  • Select from standard or long life lamps
  • Manufactured in an ISO-accredited Facility
  • Individually Tested and Issued with a Test Certificate
Our range of deuterium (D2) lamps are made to fit the majority of detectors used in analytical labs today. Our lamps are used by many of the major pharmaceutical and contract research organizations (CROs) around the globe.
Our lamps are manufactured under stringent ISO 9001 quality procedures. Each lamp is manufactured to rigorous standards and traceable from manufacturing to customer delivery.
$515.69 - $1,775.43CAD / Each
50 variations of this product are available.
See All Product Options Below
50 PRODUCT OPTIONS
Compareitem specifications
Compatible With Description
Availability Pricing
COMPARE
Agilent 8452 Deuterium (D2) Detector Lamp for Agilent 8452 Detectors
$1,421.82CAD / Each
COMPARE
Aminco DW2C, DW2000 Deuterium (D2) Detector Lamp for Aminco DW2C, DW2000
$1,237.65CAD / Each
COMPARE
Analytik Jena (Zeiss) Deuterium (D2) Detector Lamp for Analytik Jena (Zeiss)
$1,295.11CAD / Each
COMPARE
Analytik Jena Spekol Deuterium (D2) Detector Lamp for Analytik Jena Spekol
$1,370.25CAD / Each
COMPARE
Beckman DU600, 620, 630, 640, 650, 6000, 6500, 7000, 7500 Deuterium (D2) Detector Lamp for Beckman DU600, 620, 630, 640, 650, 6000, 6500, 7000, 7500
$1,242.07CAD / Each
COMPARE
Applied Biosystems 757, 759, 783A, 785A, 1000S, 980, 120A, 130A Deuterium (D2) Detector Lamp for Applied Biosystems 757, 759, 783A, 785A, 1000S, 980, 120A, 130A
$626.19CAD / Each
COMPARE
Agilent 1100 VWD, 1200, 1260 Deuterium (D2) Detector Lamp for Agilent 1100 VWD Detectors
$1,060.84CAD / Each
COMPARE
Agilent 1100 DAD, 8453 Deuterium (D2) Detector Lamp for Agilent 8453 Detectors
$996.01CAD / Each
COMPARE
Agilent 1100/1200 DAD (to fit G1315/1365 A & B series detectors) Deuterium (D2) Detector Lamp for Agilent 1100/1200 DAD (G1315/G1365 A & B Series Detectors)
$1,212.60CAD / Each
COMPARE
Agilent 1100/1200 DAD (to fit G1315/1365 C & D series detectors) Deuterium (D2) Detector Lamp for Agilent 1100/1200 DAD (G1315/G1365 C & D Series Detectors)
$1,480.76CAD / Each
COMPARE
Agilent 1260, 1290 Deuterium (D2) Detector Lamp for Agilent 1290 DAD (8-Pin) Detectors
$1,221.44CAD / Each
COMPARE
Biotage® SP1/4, Isolera Deuterium (D2) Detector Lamp for Biotage SP1/4, Isolera
$1,384.99CAD / Each
COMPARE
Dionex Ultimate 3000, 3000RS, 3100, 3400 Deuterium (D2) Detector Lamp for Dionex Ultimate VWD 3000, 3100, 3400, Longlife
$945.92CAD / Each
COMPARE
Gilson 170 DAD Deuterium (D2) Detector Lamp for Gilson 170 DAD, longlife
$599.67CAD / Each
COMPARE
Dionex/Gynkotek UVD160, 170S, 170U, 320, 340S, 340U Deuterium (D2) Detector Lamp for Dionex / Gynkotek UVD 320, 340S, 160, 170S, 170U
$1,010.75CAD / Each
COMPARE
Interchim PuriFlash (PF) 215, PF430, PF450, PF800, PF4100, PF1, SP2 Deuterium (D2) Detector Lamp for Interchim Puriflash PF215, PF430
$1,384.99CAD / Each
COMPARE
Shimadzu SPD-10A, AVP, AV, AVVP, M10AVP, 20A, 20AV, M20A Deuterium (D2) Detector Lamp for Shimadzu SPD-10A, AVP, AV, AVVP, M10AVP, 20A, 20AV
$773.53CAD / Each
COMPARE
Shimadzu spectrophotometers Deuterium (D2) Detector Lamp for Shimadzu UV Series
$515.69CAD / Each
COMPARE
Shimadzu LC2010 Deuterium (D2) Detector Lamp for Shimadzu LC2010 Longlife
$841.31CAD / Each
COMPARE
Thermo TSP UV100, 150, 200, 1000, 2000, 3000 Deuterium (D2) Detector Lamp for TSP UV100, 150, 200, 1000, 2000, 3000
$956.23CAD / Each
Showing 1 - 20 of 50