Idex® Inline Solvent MicroFilter Accessories

Use with solvent MicroFilters to protect your columns from particulates
  • Optimize performance of your solvent MicroFilters
Choose from a wide range of accessories including end-fittings, capsules, and more suitable for use with your Idex® inline solvent MicroFilter.
$33.45 - $314.20CAD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Material Compatible With Description
Availability Pricing
Idex M-500-04 MicroFilter Tubing and Ferrule Assembly for 02010-46; 1/EA RK-02010-47 Mfr # M-500-04
COMPARE
PEEK 02010-46 MicroFilter Tubing and Ferrule Assembly for 02010-46; 1/EA
$33.45CAD / Each
Idex M-120X End-Fitting for MicroTight Tubing Sleeve, Black PEEK, 0.5 µm PEEK Frit; 10/PK RK-42711-24 Mfr # M-120X
COMPARE
PEEK 42711-31 End-Fitting for MicroTight Tubing Sleeve, Black PEEK, 0.5 µm PEEK Frit; 10/PK
$314.20CAD / Pkg of 10
Idex M-133 NanoFilter Capsule, Black/Tan PEEK, 1 µm SS Frit; 2/PK RK-42711-27 Mfr # M-133
COMPARE
PEEK 42711-37 and -38 NanoFilter Capsule, Black/Tan PEEK, 1 µm SS Frit; 2/PK
$147.34CAD / Pkg of 2
Idex M-140 End-Fitting for 1/32" OD Tubing, Natural PEEK, 0.5 µm PEEK Frit; 5/PK RK-42711-30 Mfr # M-140
COMPARE
PEEK 42711-33 End-Fitting for 1/32\" OD Tubing, Natural PEEK, 0.5 µm PEEK Frit; 5/PK
$47.15CAD / Pkg of 5
Idex M-520-01 Inline MicroFilter Body, 0.006" Bore; 1/EA RK-42711-32 Mfr # M-520-01
COMPARE
PEEK 42711-29, -31, and -33 Inline MicroFilter Body, 0.006\" Bore; 1/EA
$73.67CAD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat