Cole-Parmer® VapLock™ Adapter Fittings, Compression to Threaded UNF(M), Straight
Designed for secure liquid-tight connections
  • Two piece nut and ferrule system
  • Adapter for semi-rigid tubing
  • Color coded for your convenience
Adapters for semi-rigid tubing are color coded by tubing size for easy identification. A two-piece nut and ferrule system provides a liquid-tight connection and withstands pressures up to 500 psi.
$6.78 - $109.47CAD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Compression Size / Tube Od (Port 1) (mm) Thread size UNF(M) (port 2) (InchShort)
Availability Pricing
COMPARE
White / Blue 4.76 5/16-24
$15.03CAD / Each
COMPARE
Green / Blue 1.6 1/4-28
$71.16CAD / Pkg of 10
COMPARE
Red / Green 2 1/4-28
$93.41CAD / Pkg of 10
COMPARE
Black / White 2.2 1/4-28
$50.10CAD / Pkg of 10
COMPARE
Orange / White 2.5 1/4-28
$94.59CAD / Pkg of 10
COMPARE
Yellow / Orange 3 1/4-28
$109.47CAD / Pkg of 10
COMPARE
Blue / Yellow 3.175 1/4-28
$64.24CAD / Pkg of 10
COMPARE
Red / Green 2 1/4-28
$9.58CAD / Each
COMPARE
Blue / Yellow 3.175 1/4-28
$6.78CAD / Each
COMPARE
Green / Yellow 3.175 1/4-28
$7.51CAD / Each
COMPARE
Red / Yellow 3.175 1/4-28
$7.51CAD / Each
COMPARE
Yellow / Yellow 3.175 1/4-28
$7.51CAD / Each
Showing 1- 12 of 12