Cole-Parmer® VapLock™ Adapter Fittings, Compression to Threaded NPT(M), Straight
Designed for secure leak-resistant connections
  • Easily identify from a distance with color coded adapters
Adapters for semi-rigid tubing are color coded by tubing size for easy identification. Each connection is comprised of a two-piece nut and ferrule system with polypropylene NPT(M) thread adapter. Wrap NPT thread 3 to 4 times with PTFE tape and wrench tighten for a leak-resistant connection. Adapters withstand pressures up to 500 psi.
$28.14 - $45.82CAD / Each
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Compression Size / Tube Od (Port 1) (mm) Thread Size NPT(M) (Port 2) (InchShort)
Availability Pricing
COMPARE
Brown nut/natural ferrule/white adapter 4 1/4
$45.82CAD / Each
COMPARE
White nut/blue ferrule/white adapter 4.76 1/4
$40.08CAD / Each
COMPARE
Green nut/blue ferrule/white adapter 1.6 1/4
$28.14CAD / Each
COMPARE
Red nut/green ferrule/white adapter 2 1/4
$29.17CAD / Each
COMPARE
Black nut/white ferrule/white adapter 2.2 1/4
$28.29CAD / Each
COMPARE
Orange nut/white ferrule/white adapter 2.5 1/4
$28.29CAD / Each
COMPARE
Yellow nut/orange ferrule/white adapter 3 1/4
$30.20CAD / Each
COMPARE
Blue nut/ yellow ferrule/white adapter 3.175 1/4
$28.29CAD / Each
COMPARE
Green nut/ blue ferrule/white adapter 1.6 1/8
$33.74CAD / Each
COMPARE
Red nut/green ferrule/white adapter 2 1/8
$33.59CAD / Each
COMPARE
Black nut/white ferrule/white adapter 2.2 1/8
$33.89CAD / Each
COMPARE
Orange nut/natural ferrule/white adapter 2.5 1/8
$34.04CAD / Each
COMPARE
Yellow nut/orange ferrule/white adapter 3 1/8
$33.59CAD / Each
COMPARE
Blue nut/ yellow ferrule/white adapter 3.175 1/8
$36.39CAD / Each
Showing 1- 14 of 14