Cole-Parmer Fast-Response Autoclavable Fermentation pH Electrodes
New glass formulation provides unparalled performance!
  • Lightening fast response time with superior accuracy and repeatabilty
  • Perfect for low-ionic strength solutions
  • Provides confident values over the entire pH range from 0 to 14
Cole-Parmer's fast-response fermentation pH electrodes are made of a proprietary new formulation of low-impedance glass replaces all other formulas — allows you to streamlined inventories and eliminate wasted time trying to select the best electrode for your application. Use in low-ionic strength solutions, high pH testings where sodium error could be a problem, and at high temperatures up to 266°F (130°C). pH is registered in less than 20 seconds, with a typically accuracy of 0.02 pH units (meter dependant).
Chemical-resistant Ryton ''PPS'' PG13.5 threaded pressure fitting, PTFE spacer, and silicone o-ring ensure a leakproof connection to your assembly.
All electrodes feature double-junction reference for maximum lifespan.
Order detachable cap
$134.08 - $515.69CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
pH Electrode, Fermentation, 12 x 225 mm, annular junction, ATC, 4-pin connection
$515.69CAD / Each
COMPARE
pH Electrode, Fermentation, 12 x 120 mm, annular junction, S8 Connection
$387.50CAD / Each
COMPARE
pH Electrode, Fermentation, 12 x 225 mm, Open junction, S8 connection
$431.70CAD / Each
COMPARE
pH Electrode, Fermentation, 12 x 120 mm, Open junction, S8 connection
$374.24CAD / Each
COMPARE
Cable, 4-pin to BNC w/ATC wires for 29045 series pH electrodes, 3-ft
$134.08CAD / Each
COMPARE
Cable, S8 to BNC for 29045 series pH electrodes, 3-ft
$137.76CAD / Each