Trajan SGE Replacement Plunger, for 5 mL Removable Needle Gas Tight Syringe
Representative image only.
Trajan SGE Replacement Plunger, for 5 mL Removable Needle Gas Tight Syringe
Trajan | Mfr: 031856| Item: 1600269
Choose wisely to save your operational cost—no need to buy new syringes instead
$120.52CAD / Pkg of 1
$120.52CAD / Pkg of 1
Buy It Together
Current Item
Trajan SGE Replacement Plunger, for 5 mL Removable Needle Gas Tight Syringe
Trajan SGE Replacement Plunger, for 5 mL Removable Needle Gas Tight Syringe
1600269
$120.52CAD / Pkg of 1
+ =
Total Price 325.69 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Each stainless steel plunger is individually fitted to each barrel
  • 6 month warranty
More About this Item
All SGE gas-tight syringes have replaceable plungers.
The standard plunger in MicroVolume syringes is not replaceable.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Replacement plunger, for 5 mL Removable Needle Gas Tight Syringe; 1/Ea