TPI 367DA367 Waterproof Themocouple and Thermistor Datalogger Thermometer
Representative image only.
TPI 367DA367 Waterproof Themocouple and Thermistor Datalogger Thermometer
TPI | Mfr: 367DA367| Item: UZ-96001-80
All metal housing make this meter extremely rugged, but still easy to use
$462.64CAD / Each
for an additional
$312.73CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$462.64CAD / Each
Buy It Together
Current Item
TPI 367DA367 Waterproof Themocouple and Thermistor Datalogger Thermometer
TPI 367DA367 Waterproof Themocouple and Thermistor Datalogger Thermometer
9600180
$462.64CAD / Each
+ + =
Total Price 929.56 ADD 3 ITEMS TO CART
Product Description
Includes
Users manual and 2 AA batteries.
Key Features
  • Auto field calibration
  • Data logging capability: over 10,000 readings
  • Real time and logged data transfer to PC via USB docking station
  • Fast reading mode - Increases sample rate
  • Multi use belt clip which can used as a belt clip or wall mount
  • 3 year warranty
More About this Item
The A367D is designed for harsh environments featuring water tight construction and uses Lumberg connection probes to ensure that water and dirt do not infiltrate into the meter. A rugged metal housing with over molded boot further protects the instrument insuring a long life. The meter can use both thermocouple and thermistor probes and can datalog over 10,000 readings at intervals from 1 second to 24 hours. Stored data is transfers to docking station through an infrared port on the back of the meter, the data can then be uploaded to a PC via a USB cable. The unit can also be field calibrated, through an easy 2-step process performed with the keypad. Other features include, two line LCD display, clock with alarm, alarm timer, over temperature alarm, Min/Max, record, data hold, °C/°F selectable, and auto off.
3 Year Warranty
Specifications
  • Meter Measurement TypeMulti-Parameter
  • Number Of Probe Channels1
  • Types Of Probes AcceptedType J, Type K
  • Min Temperature (° F)-328
  • Max Temperature (° F)2498
  • Min Temperature (° C)-200
  • Max Temperature (° C)1112
  • Accuracy±1% of reading
  • Resolution0.1°/1° (user selectable)
  • OutputUSB
  • Data LoggingYes
  • Memory Capacity16000 data points
  • IP RatingIP67
  • Low Battery IndicatorYes
  • Display Type4.5-digit LCD
  • Battery Life7000 hours
  • Battery2 AA (included)
  • DescriptionWaterproof Themocouple and Thermistor Datalogger Thermometer