Cole-Parmer SamplePrep HG-250 GenoLyte® Temperature Controlled Compact Tissue Homogenizer and Cell Lyser; 115/230 VAC, 50/60 Hz

Representative image only.

Cole-Parmer SamplePrep HG-250 GenoLyte® Temperature Controlled Compact Tissue Homogenizer and Cell Lyser; 115/230 VAC, 50/60 Hz

Cole-Parmer SamplePrep | Mfr: 1200C| Item: 0450005

Ideal for small-scale sample preparation

Call For Price

Call For Price

Buy It Together

Current Item

Cole-Parmer SamplePrep HG-250 GenoLyte® Temperature Controlled Compact Tissue Homogenizer and Cell Lyser; 115/230 VAC, 50/60 Hz
Cole-Parmer SamplePrep HG-250 GenoLyte® Temperature Controlled Compact Tissue Homogenizer and Cell Lyser; 115/230 VAC, 50/60 Hz
0450005

Call For Price
+ =
Total Price 508.32 ADD 2 ITEMS TO CART
Product Description

Includes

Vial holder for 6 x 2mL vials. Additional size holders must be ordered separately.

Key Features

  • High-intensity oscillating motion homogenizes samples in seconds
  • Keep an eye on the grind process with the clear lid for viewing
  • Vial holder for 6 x 2 mL vials (included)
  • Interchangeable vial holders handle a range of vial sizes from 2, 5, 7, 12 and 15 mL (sold separately)
  • Safety interlock switch stops the motor if the lid is opened during homogenization
  • Digital display screen with rate and timer controls allows you to adjust speed and set time
  • Clear lid allows you to see the grinding process
  • Quick-connect to a cooling system
  • Sample temperature maintained between 0-10 ºC
  • Preserve RNA and protein
  • Cool samples without freezing
  • Maintain sample temperature throughout the run
  • High-intensity oscillating motion homogenizes samples in seconds
  • Accommodate and holds up to three 2 mL standard vials
  • Programmable control panel
  • Three fixed speed options
  • Lid has a safety interlock switch that stops the motor if the lid is opened during the grinding process

More About this Item

The GenoLyte is capable of blending powders and crushing rocks to a fine powder or DNA, RNA, and protein extractions. It is specifically designed for rapid cell disruption, cell lysis and tissue homogenization. Common samples include plant material, seeds, animal tissue, bacteria, yeast, stool, and soil. This homogenizer is compact yet powerful, which frees up bench space in the lab. Homogenizer accepts a vial holder for 2 to 12 mL vials (sold separately).

The GenoLyte® is a powerful, compact, temperature controlled homogenizer. Ideal for applications involving sample preparation for DNA/RNA research and extraction, protein extractions, tissue homogenization, and cell lysis.

The HG-250 Chilled GenoLyte® is an efficient, compact laboratory homogenizer that uses a circulating-chiller (not included) to maintain sample temperature between 0 to 10°C while effectively disrupting cellular materials by horizontally oscillating 2 mL vials.

Typical applications include: DNA/RNA research and extraction, protein extractions, and tissue homogenization.

Typical samples include: animal tissue, plant tissue, cell cultures, bacteria, yeast, stool, and soil.

The HG-250 Chilled GenoLyte® is a compact, high-performance laboratory homogenizer designed to keep samples cool while ensuring efficient, consistent tissue disruption. By maintaining stable temperatures between 0 to 10 °C (when connected to a circulating chiller, not included), while effectively disrupting cellular materials by horizontally oscillating 2 mL vials, this system prevents heat-related degradation, making it ideal for sensitive nucleic acid, protein, and enzyme extractions.

The Cole-Parmer SamplePrep HG-250 Temperature Controlled GenoLyte maintains stable low temperatures to protect heat-sensitive molecules, preventing sample degradation during processing.

The unit effectively disrupts tough samples like seeds, stems, roots, leaves, and animal tissue in minutes using bead beating technology. This technology quickly extracts nucleic acids, proteins, and other biomolecules.

The quick-connect cooling setup makes it easy to integrate a chiller. The digital timer ensure precise control over tune time and speed. The lid includes a safety interlock for added protection, and the clear lid design allows for visibility of the grinding process. Clamp orientation makes loading and unloading the vial effortless and ensures perfect vial placement.

The mechanical disruption eliminates the time-consuming and inconsistent process of manual grinding, ensuring reproducible extractions without the risk of cross-contamination.

Operation is easy, simple secure the vials in the cooled sample chamber and close the lid. Set the desired time and speed using the digital controls, press start, and the the GenoLyte efficiently process your samples.

A circulating chiller is required for continuous temperature control. The GenoLyte is compatible with Polystat® Circulating Baths or most circulating chiller systems. Purchase chiller and vials separately.

Effective September 2023, the Spex SamplePrep 1200C GenoLyte® has now been rebranded Cole-Parmer® SamplePrep under model number HG-250 Temperature Controlled GenoLyte®. All the same great features and specifications, just with a new and improved look.

Made in the U.S.A.
Specifications
  • Min Sample Size (mL)2
  • Max Sample Size (mL)12
  • Max Speed (rpm)4000
  • Min Speed (rpm)750
  • Height (in)11
  • Width (in)8
  • Length (in)15
  • Height (cm)30
  • Width (cm)20.5
  • Length (cm)38
  • Power (VAC)115 / 230
  • Power (Hz)50/60
  • Motor hP0.14
  • DescriptionGenoLyte® Compact Homogenizer with 6 x 2 mL Vial Holder, 2 to 12 mL; 115/230 VAC, 50/60Hz
Reviews
Q&A