Environmental Express Oakton ORPTestr® 50 Waterproof Pocket ORP Tester, Premium 50 Series
Representative image only.
Environmental Express Oakton ORPTestr® 50 Waterproof Pocket ORP Tester, Premium 50 Series
Premium multi parameter pocket testers for field and lab testing
$532.63CAD / Each
$532.63CAD / Each
Buy It Together
Current Item
Product Description
Includes
4 AAA batteries.
Key Features
  • Available in pH, Conductivity, TDS, Salinity, and ORP models
  • Replaceable, double-junction sensors for durability and longevity against aggressive samples
  • Four-button keypad for technician level use
  • Leakproof cap functions as a sample cup, hydrating cup, and stabilizing base to stand the meter upright
  • 1 year warranty
More About this Item
Our most advanced line of pocket testers, the Oakton 50 Series provides single and multimeter options for measuring pH, Conductivity, TDS, Salinity, and ORP. Replaceable double junction sensors provide durability and longevity against highly caustic, volatile or hazardous samples in both lab and field applications. Waterproof and designed to float in water, they are ideal for field-based testing environments – with a convenient loop for security line attachment.
50 series pocket meters include a replaceable double-junction electrode sensor that is designed for more aggressive applications, including wastewater and solutions containing sulfides, heavy metals, and TRIS. The electrode sensor offers a clog-resistant Kynar® junction and a recessed, glass pH bulb to protect against breakage and allow for easy maintenance and replacement. A Valox sensor housing resists most chemicals.
An easy-to-use four-button keypad allows for technician level use and measurement with quick access to a variety of functions. Up to 3 calibration points with auto-buffer recognition at both USA and NIST standard buffers. Additional features include an automatic temperature compensation (ATC), hold function, auto-off function, self-diagnostic error messages, and a three-segment battery-life indicator.
1 Year WarrantyConformite EuropeenneIP 67ISO 9001:2008 Certified Supplier
Specifications
  • Meter TypeTester
  • Display typeLCD
  • Calibration±200 mV offset adjustment
  • Min mV (mV)-1000
  • Max mV (mV)1000
  • mV Resolution1 mV
  • mV Accuracy±2 mV
  • Min Temperature (° C)0
  • Max Temperature (° C)60
  • Min Temperature (° F)32
  • Max Temperature (° F)140
  • WaterproofYes
  • Battery4 AAA (included)
  • Battery life (hr)250
  • DescriptionWaterproof Pocket ORP Tester, Premium 50 Series