The product you searched is discontinued. Below is the suggested replacement. Compare to discontinued item.

Environmental Express DO 2000 Benchtop Meter Kit

Representative image only.

Environmental Express DO 2000 Benchtop Meter Kit

Environmental Express | Item: 3541974

Developed using extensive feedback from laboratory staff, our new Environmental Express 2000 series meters are assembled in various configurations for measuring pH/mV, EC, Ion, DO, BOD parameters

$3,144.21CAD / Each

$3,144.21CAD / Each

Buy It Together

Current Item

Environmental Express DO 2000 Benchtop Meter Kit
Environmental Express DO 2000 Benchtop Meter Kit
3541974

$3,144.21CAD / Each
+ +
Environmental Express 2000 All-in-One pH Electrode
3541952

$282.89CAD / Each
+
Environmental Express pH 2000 Benchtop Meter Kit
3541954

$1,701.77CAD / Each
+ +
Environmental Express 2000 Replacement DO Sensor
3541975

$1,444.66CAD / Each
=
Total Price 9051.18 ADD 6 ITEMS TO CART
Product Description

Key Features

  • Various configurations available to meet the demands of measuring pH/mV, EC, Ion, PC, PD, DO, BOD parameters
  • Meter Kit formats include probes, 35419-54, -60, -65, -74, -78
  • Automatic record retention and accessibility for continuous compliance with regulatory requirements
  • 0.01, 0.001 resolution for routine or research applications where acute accuracy is critical
  • Large displays with a small footprint, ideal for labs with limited bench space
  • Automatic functions like buffer recognition and auto calibration for ease of use at any skill level

More About this Item

Environmental Express provides a diverse range of water quality analyzers and electrodes that are designed specifically for laboratory applications with compliance in mind. Our multi-parameter benchtops include functionality to meet the highly specific and/or broad ranging needs of wastewater, drinking water, food and beverage, chemical and pharmaceutical testing labs.

These benchtop meters offer a high-capacity internal memory that can capture and store 2000 data points with the time and date. They have a time-based auto data log with GLP compliant real time clocks. This data can be exported into CSV, Excel, or PDF formats via USB cable and use of complimentary software. A password access protection function secures the meter setup mode from unauthorized access. An adjustable shutoff time up to 30 minutes turns the meter off when idle.

Conformite Europeenne
Specifications
  • Meter TypeDO/BOD/Temp (°C/°F)
  • Display type5” Custom LCD with backlight and 320 segments
  • ConnectionsBNC, phono (ATC), DC sockets
  • Dissolved Oxygen Resolution0.01 mg/L, 0.1%
  • Dissolved Oxygen Accuracy+/- 0.1 mg/L , +/- 1.0%
  • Min Temperature (° C)-30
  • Max Temperature (° C)130
  • Min Temperature (° F)-22
  • Max Temperature (° F)266
  • Temperature Resolution0.1 °C / °F
  • Temperature Accuracy± 0.5 °C / ± 0.9 °F
  • OutputPhono jack (USB/RS-232C)
  • Data Logging (points)2000
  • Power (VAC)100 to 240
  • Power (Hz)50-60 Hz
  • Length (cm)15.5
  • Width (cm)15
  • Height (cm)6.7
  • Width (in)5 29/32
  • Length (in)6 7/64
  • Height (in)2 41/64
  • DescriptionDO 2000 Benchtop Meter Kit
  • Warranty3 years
Reviews
Q&A