Fytertech Spilfyter Wastewater Reference Chart
Representative image only.
Fytertech Spilfyter Wastewater Reference Chart
Fytertech | Mfr: 588888| Item: 8642356
Classify unknown spills in 30 seconds
$48.18CAD / Each
$48.18CAD / Each
Buy It Together
Current Item
Fytertech Spilfyter Wastewater Reference Chart
Fytertech Spilfyter Wastewater Reference Chart
8642356
$48.18CAD / Each
+ =
Total Price 647.11 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
Convenient strips change color to identify risk type and classify spill for proper treatment. Compare color indicator to laminated, reusable reference chart to classify unknown liquid. Chemical classifier kit tests for acid or base, iodine, oxidizers, chlorine, fluoride, bromine and petroleum/-organic products. Wastewater classifier kit tests for acid/base, petroleum/organic solvent, hydrogen sulfide, nitrite, nitrate, and fluoride risks.
Replacement reference chart for Wastewater Classifier Kit.
Specifications
  • DescriptionWastewater Reference Chart