Fytertech Spilfyter Wastewater Classifier Strips, 10/Pack
Representative image only.
Fytertech Spilfyter Wastewater Classifier Strips, 10/Pack
Fytertech | Mfr: 580010| Item: 8642417
Classify unknown spills in 30 seconds
$275.89CAD / Pkg of 10
$275.89CAD / Pkg of 10
Buy It Together
Current Item
Fytertech Spilfyter Wastewater Classifier Strips, 10/Pack
Fytertech Spilfyter Wastewater Classifier Strips, 10/Pack
8642417
$275.89CAD / Pkg of 10
+ =
Total Price 874.82 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
Convenient strips change color to identify risk type and classify spill for proper treatment. Compare color indicator to laminated, reusable reference chart to classify unknown liquid. Chemical classifier kit tests for acid or base, iodine, oxidizers, chlorine, fluoride, bromine and petroleum/-organic products. Wastewater classifier kit tests for acid/base, petroleum/organic solvent, hydrogen sulfide, nitrite, nitrate, and fluoride risks.
Specifications
  • DescriptionWastewater Classifier Strips, 10/Pack