Fytertech Spilfyter Chemical test strips, case of 50
Representative image only.
Fytertech Spilfyter Chemical test strips, case of 50
Fytertech | Mfr: 570050| Item: 8642350
Classify unknown spills in 30 seconds
$1,395.30CAD / Case of 50
$1,395.30CAD / Case of 50
Buy It Together
Current Item
Fytertech Spilfyter Chemical test strips, case of 50
Fytertech Spilfyter Chemical test strips, case of 50
8642350
$1,395.30CAD / Case of 50
+ =
Total Price 1994.23 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
Convenient strips change color to identify risk type and classify spill for proper treatment. Compare color indicator to laminated, reusable reference chart to classify unknown liquid. Chemical classifier kit tests for acid or base, iodine, oxidizers, chlorine, fluoride, bromine and petroleum/-organic products. Wastewater classifier kit tests for acid/base, petroleum/organic solvent, hydrogen sulfide, nitrite, nitrate, and fluoride risks.
Replacement test strips for Chemical Classifier Kit.
Specifications
  • DescriptionChemical test strips, case of 50