Labconco FilterMate™ Ammonia Carbon Filter for FilterMate™ Portable Exhausters
Representative image only.
Labconco FilterMate™ Ammonia Carbon Filter for FilterMate™ Portable Exhausters
Labconco | Mfr: 3923402| Item: UZ-09101-54
Filter removes contaminants before returning air to room or exhausting to outside
$1,163.98CAD / Each
$1,163.98CAD / Each
Product Description
Key Features
  • Compatible with Labconco® FilterMate™ portable exhausters
  • Type CX
  • 16 lb of impregrated carbon media
More About this Item
Optimize performance of your Labconco FilterMate™ portable exhausters with appropriate filters.
Treated carbon media is suitable for ammonia and low molecular weight amines.
Specifications
  • Filter TypeAmmonia carbon
  • DescriptionAmmonia Carbon Filter for FilterMate™ Portable Exhausters