Idex Port Plug, 5/16" Hex Head, Natural ETFE, 1/4-28 Flat-Bottom; 1/EA
Representative image only.
Idex Port Plug, 5/16" Hex Head, Natural ETFE, 1/4-28 Flat-Bottom; 1/EA
Idex | Mfr: P-123| Item: UZ-02018-33
Available in various head styles to suit your application
$4.27CAD / Each
$4.27CAD / Each
Product Description
Key Features
  • Choose from ETFE or PEEK construction
  • 5/16" hex head
  • ETFE construction
  • 1 year warranty
More About this Item
Use the plugs to close off unused ports in valves and multiport connectors.
1 Year Warranty
Specifications
  • MaterialETFE
  • ColorNatural
  • Compatible With1/4-28 flat-bottom port
  • DescriptionPort Plug, 5/16" Hex Head, Natural ETFE, 1/4-28 Flat-Bottom; 1/EA