HDPE Faucet with 3/4" NPT(M) Thread, 5/Pk
Representative image only.
HDPE Faucet with 3/4" NPT(M) Thread, 5/Pk
| Item: 0607800
Instant on/off control ensures fast, smooth flow
$107.41CAD / Pkg of 5
BUY MORE & SAVE
  • 1-5$107.41 CAD / Pkg of 5
  • 6-29$96.67 CAD / Pkg of 5
  • 30-119$91.30 CAD / Pkg of 5
  • 120+$85.93 CAD / Pkg of 5
$107.41CAD / Pkg of 5
BUY MORE & SAVE
  • 1-5$107.41 CAD / Pkg of 5
  • 6-29$96.67 CAD / Pkg of 5
  • 30-119$91.30 CAD / Pkg of 5
  • 120+$85.93 CAD / Pkg of 5
Product Description
Key Features
  • Stem is LDPE
  • 1 year warranty
More About this Item
These faucets are manufactured of HDPE for greater strength and positive thread engagement.
The stem is LDPE for 06078-00.
1 Year Warranty
Specifications
  • MaterialHigh Density Polyethyene (HDPE)
  • Cap MaterialHigh-Density Polyethyelene (HDPE)
  • Cap / Stopper Size3/4" NPT
  • Cap ColorBrown
  • AutoclavableNo
  • DescriptionHDPE Faucet with 3/4" NPT(M) Thread, 5/Pk