Hach 5-EP Hardness Drop Test Kit -Total and Calcium; 1 to 30 ppm Caco3
Representative image only.
Hach 5-EP Hardness Drop Test Kit -Total and Calcium; 1 to 30 ppm Caco3
Hach | Mfr: 145400| Item: UZ-05545-32
Low cost and simple drop count titration method
$85.60CAD / Each
$85.60CAD / Each
Buy It Together
Current Item
Hach 5-EP Hardness Drop Test Kit -Total and Calcium; 1 to 30 ppm Caco3
Hach 5-EP Hardness Drop Test Kit -Total and Calcium; 1 to 30 ppm Caco3
0554532
$85.60CAD / Each
+ + =
Total Price 1198.16 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Kit contains all required reagents and apparatus
  • 1 year warranty
More About this Item
Drop count titration is a simple method to estimate concentrations.
Rugged carrying case for analysis in the field
ISO9001 Certified Supplier1 Year Warranty
Specifications
  • TestHardness, total
  • Measurement Points1 to 30 ppm
  • Reagent PlatformEDTA Powder Pillows
  • Tests Per Kit100
  • DescriptionHardness Drop Test Kit -Total and Calcium; 1 to 30 ppm Caco3