Hach 21056-69 Colorimeter Test Kit , Total Chlorine (USEPA), 0 to 2.0 ppm, 100/Kit
Representative image only.
Hach 21056-69 Colorimeter Test Kit , Total Chlorine (USEPA), 0 to 2.0 ppm, 100/Kit
Hach | Mfr: 2105669| Item: UZ-99574-05
Environmental test kits for your Hach Colorimeter
$48.84CAD / Each
$48.84CAD / Each
Buy It Together
Current Item
Hach 21056-69 Colorimeter Test Kit , Total Chlorine (Usepa), 0 to 2.0 ppm, 100/Kit
Hach 21056-69 Colorimeter Test Kit , Total Chlorine (Usepa), 0 to 2.0 ppm, 100/Kit
9957405
$48.84CAD / Each
+ + =
Total Price 1210.09 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 1 year warranty
More About this Item
Order the test kits for the Hach colorimeter to complete your water testing protocol. All kits require a colorimeter (99573-00 shown below). Chemical oxygen demand (COD), Total Nitrogen and Totol Phosphorus require a COD reactor (99574-60, 120 VAC or 99574-65, 240 VAC).
1 Year Warranty
Specifications
  • Test ParametersTotal Chlorine (USEPA)
  • Number of Tests Included100
  • Range0 to 2.0 ppm
  • DescriptionColorimeter Test Kit , Total Chlorine (Usepa), 0 to 2.0 ppm, 100/Kit