Fluke FLK-T3000 FC Wireless Type K Thermocouple Module

Representative image only.

Fluke FLK-T3000 FC Wireless Type K Thermocouple Module

Fluke | Mfr: FLK-T3000 FC| Item: UZ-20047-79

See, save, and share data wirelessly — saving time and increasing productivity

$449.37CAD / Each

for an additional
$312.73CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$449.37CAD / Each

Buy It Together

Current Item

Fluke FLK-T3000 FC Wireless Type K Thermocouple Module
Fluke FLK-T3000 FC Wireless Type K Thermocouple Module
2004779

$449.37CAD / Each
+ =
Total Price 881.81 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Increase productivity by communicating wirelessly between your phone, PC, and Fluke test tools
  • Download the Fluke Connect™ app for use with iOS (4s and later) and Android™ to connect with over twenty Fluke test tools (Download iOS app at the App Store, and the Android app from Google play)
  • Logging function on modules allow you to isolate intermittent events or record signal fluctuations remotely
  • Share data with associates in seconds, allowing for immediate interpretation and action
  • Increase efficiency by simultaneously viewing multiple measurements on one screen
  • Take measurements from safe locations reducing potential hazards
  • 3 year warranty

More About this Item

The Fluke T3000 FC K-type thermocouple thermometer uses the 80PK-1 K-type bead thermocouple for taking temperature measurements and can be used as a standalone meter or as part of the Fluke Connect wireless system. The logging function records and saves up to 65,000 readings for added flexibility.

The Fluke T3000 FC is a versatile K-type thermocouple thermometer that wirelessly relays measurements to other Fluke Connect enabled master units listed below:
Fluke 3000 FC DMM
Fluke Ti200/300/400 Infrared Cameras
To your PC via optional PC3000 FC adapter

What’s included: K-type temperature bead probe, magnetic hanging strap, and two AA batteries.

Available for sale in the United States and Canada only

3 Year WarrantyConformite EuropeenneCanadian Standards Association
Specifications
  • Min Temperature (° F)-328
  • Max Temperature (° F)2501
  • Min Temperature (° C)-200
  • Max Temperature (° C)1372
  • Temperature Accuracy±0.5% + 0.3 counts
  • Width (in)2.5
  • Height (in)6.5
  • Depth (in)1.25
  • Width (cm)6.35
  • Height (cm)16.5
  • Depth (cm)3.75
  • DescriptionWireless Type K Thermocouple Module
  • Warranty3 Years
Reviews
Q&A