Digi-Sense Heavy-Duty Industrial Digital Multimeter with Bluetooth® Connectivity

Representative image only.

Digi-Sense Heavy-Duty Industrial Digital Multimeter with Bluetooth® Connectivity

Digi-Sense | Item: 2025064

View, store, and share real-time data from a safe distance on your smartphone

$306.47CAD / Each

$306.47CAD / Each
Product Description

Includes

Test leads, type K thermocouple probe with mini connector, carrying case, 9 V battery, and NIST-traceable calibration.

Key Features

  • Get the most out of your data with our user-friendly mobile app
  • Unlimited data storage based on smart device capacity
  • Reliable measurements, guaranteed! Precalibrated to NIST-traceable standards
  • Rugged double-molded housing provides increased durability
  • Bright, large backlit 4000-count LCD permits viewing in low-light conditions
  • Tilt stand offers easy hands-free viewing
  • IP67 waterproof rated
  • CAT IV-600 V, CAT III-1000 V safety rating
  • 2 year warranty

More About this Item

Using our free Digi-Sense D/S Meter app, this heavy-duty industrial digital multimeter with Bluetooth® technology allows you to transmit data directly to your Android or iOS device. Now you can view data at a safe distance from dangerous equipment or hazardous locations and even use your smart device as a real-time secondary display for checking measurements, eliminating the need to write down readings on paper. The results are automatically stored in the app with a date-and-time stamp and can be saved as a CSV file and emailed for future reference, manipulation, or analysis to help determine trends and conditions over a long period of time. Use the app to attach photos and notes to the records creating a clear reference point for your work—ideal for record keeping at large sites.

The handheld digital multimeter measures AC/DC voltage, AC/DC current, resistance, frequency, capacitance, and continuity. Special functions include auto ranging, data Hold, Max/Min readings, diode test, audible continuity check, relative mode, duty cycle, low-battery indication, and auto power-off. The meter also offers True RMS for more accurate current readings, temperature measurement with a type K thermocouple, and analog bar graph for viewing trends.

Two year warrantyApple App StoreConformite EuropeenneGoogle Play AppIP 67
Specifications
  • DC Min (Volts)0.4
  • DC Max (Volts)1000
  • DC Voltage Accuracy±1.0% rdg + 8 digits
  • DC Min (amps)0.0004
  • DC Max (amps)10
  • DC Current Accuracy±1.0% rdg + 3 digits
  • AC Min (Volts)0.4
  • AC Max (Volts)1000
  • Ac Voltage Accuracy±1.5% rdg + 10 digits
  • AC Min (amps)0.0004
  • AC Max (amps)10
  • AC Current Accuracy±3.0% rdg + 5 digits
  • Resistance Max (Ω)400
  • Resistance Min (MΩ)40
  • Capacitance Min (nF)4000000
  • Capacitance Max (nF)40
  • Capacitance Accuracy±3.0% rdg ± 5 digits
  • Frequency Max (Hz)9.999
  • Frequency Min (MHz)9.999
  • Frequency Accuracy±1.5% ± 5 digits
  • Resistivity Accuracy±1.2% rdg ± 4 digits
  • ContinuityYes
  • Diode TesterYes
  • Duty CycleYes
  • True RmsYes
  • Min Temperature (° F)-50
  • Max Temperature (° F)1382
  • Min Temperature (° C)-45
  • Max Temperature (° C)750
  • Temperature Accuracy±3.0% rdg ± 9°F (5°C)
  • DisplayBacklit LCD
  • MemoryUnlimited data storage based on smart device capacity
  • Safety RatingCAT IV-600 V, CAT III-1000 V
  • PowerOne 9 V battery
  • Width (in)3.25
  • Length (in)2.25
  • Height (in)7
  • Width (cm)8.2
  • Length (cm)5.7
  • Height (cm)18
  • DescriptionHeavy Duty Industrial Digital Multimeter with Bluetooth
  • Warranty2 year
Reviews
Q&A