Delmhorst JLX-30 Leather Moisture Meter
Representative image only.
Delmhorst JLX-30 Leather Moisture Meter
Delmhorst | Mfr: JLX-30W/CS| Item: UZ-59800-86
The JLX-30 is the go-to tool for tanneries and leather goods manufacturers, providing professional-grade moisture monitoring to ensure high quality and consistency in every product
$1,075.57CAD / Each
$1,075.57CAD / Each
Product Description
Includes
Meter and carrying case.
Key Features
  • Moisture range from 10-28% for leather
  • Large, easy-to-read display with auto-backlight and on-screen statistics for up to 100 readings
  • Bluetooth® connectivity to the EDGE® app for data export, LED customization, and more
  • Customizable alarm set point and color-coded LED alerts for optimal moisture control
  • Sturdy ABS case with ergonomic E-Z grip handle and integrated contact pins
  • 2 year warranty
More About this Item
Built for durability and precision, the JLX-30 Leather Moisture Meter provides reliable moisture readings across production stages, including drying and quality control. With Bluetooth® connectivity, this advanced tool links to the free EDGE® app, allowing users to timestamp and geotag readings, export data, and customize LED indicators. The rugged ABS case, along with a user-friendly dashboard and customizable settings, makes the JLX-30 an indispensable tool for leather professionals dedicated to superior quality and long-lasting reliability.
Specifications
  • Measurement TypeLeather (10-28%)
  • Temperature CompensationYes
  • Min Temperature (° F)0
  • Min Temperature (° C)-18
  • Max Temperature (° F)255
  • Max Temperature (° C)124
  • Display TypeLED
  • Data LoggingYes
  • Memory Capacity100 readings
  • AlarmUser-selected MC level
  • OutputBluetooth®
  • Battery2 AA
  • Electrode TypeNone included
  • Height (in)1 19/32
  • Width (in)2 27/32
  • Length (in)8 21/32
  • DescriptionLeather Moisture Meter
  • Warranty2 years