Environmental Express pH electrode, Refillable, long and thin body
Representative image only.
Environmental Express pH electrode, Refillable, long and thin body
Environmental Express | Item: 0599161
Faster response and increased stability across a pH range of 0 to 14
$361.28CAD / Each
BUY MORE & SAVE
  • 1-3$361.28 CAD / Each
  • 4+$325.15 CAD / Each
$361.28CAD / Each
BUY MORE & SAVE
  • 1-3$361.28 CAD / Each
  • 4+$325.15 CAD / Each
Buy It Together
Current Item
Environmental Express pH electrode, Refillable, long and thin body
Environmental Express pH electrode, Refillable, long and thin body
0599161
$361.28CAD / Each
+ + =
Total Price 685.94 ADD 3 ITEMS TO CART
Product Description
Key Features
  • Dome bulb offers better protection from breakage
  • Stability in low-ionic strength solutions
  • Offers resistance to Na+ ion interference
  • Compatible with most brands of pH meters (that accept standard BNC connections)
  • 6 month warranty
More About this Item
Glass body combination probes are able to withstand higher temperatures and chemical attack. All have 3.3 ft (1 m) long cables with BNC connectors.
6 Month Warranty
Specifications
  • ApplicationsSmall test tubes sampling
  • Electrode TypeSingle junction
  • Electrode Body MaterialGlass
  • Reference ElectrolyteAg/AgCl
  • Max Temperature (° C)100
  • ConnectorBNC
  • DescriptionpH electrode, Refillable, long and thin body
  • Warranty6 month warranty