The product you searched is discontinued. Below is the suggested replacement. Compare to discontinued item.

Environmental Express PC 100 Portable pH/Conductivity Meter with pH, Conductivity, and ATC Probes

Representative image only.

Environmental Express PC 100 Portable pH/Conductivity Meter with pH, Conductivity, and ATC Probes

Environmental Express | Item: 3561345

Measure pH, conductivity, and temperature at the same time!

$1,166.92CAD / Each

$1,166.92CAD / Each
Product Description

Includes

Oakton PC100 Portable pH/Conductivity Meter, pH and conductivity probes, electrode clip, temperature probes, and 3 AA batteries.

Key Features

  • Measured parameter and temperature shown on display at same time
  • Convenient, compact design with large display
  • Stability, calibration, and low-battery indicators
  • Waterproof and dust-resistant IP57 rating – great for use under harsh environments
  • 2 year warranty

More About this Item

The new addition to the Environmental Express Oakton product line features updates to measuring water quality easier and faster. The larger display is in. by 2.0 in. for better viewing angle, and has indicators for battery life, stability, and calibration. It also shows both the parameter and temperature measurements at the same time. The PC 100 meter measures pH, mV, conductivity, TDS, salinity, and temperature.

The built-in microprocessor chip enables advanced functions, such as auto calibration, auto temperature compensation, parameter setting, self-diagnosis, auto power-off, and low-battery reminders. The meter uses advanced digital processing technology, intelligently improving the response time and accuracy of your measurements. Stable reading and auto lock display modes are available.

Two year warrantyConformite Europeenne
Specifications
  • Meter TypepH, mV, ORP, conductivity, TDS, salinity, temperature
  • Display typeLCD
  • ConnectionsBNC & Φ2.5 socket
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.01/0.1 pH
  • pH Accuracy±0.01 pH
  • Buffer RecognitionAutomatic recognition of USA, NIST, and user defined pH calibration solutions; Automatic recognition of standard and user-defined conductivity calibration solutions
  • CalibrationpH: 1-3 points auto calibration; Conductivity: 1-4 points automatic
  • Min mV (mV)-1000
  • Max mV (mV)1000
  • mV Resolution0.1 mV from –200 to 200 mV; 1 mV remaining
  • mV Accuracy±0.2% full-scale
  • Min Conductivity0 mS/cm
  • Max Conductivity0.00 to 19.99 μS/cm, 20.0 to 199.9 μS/cm, 200 to 1999 μS/cm; 2.00 to 19.99 mS/cm, 20.0 to 199.9 mS/cm
  • Conductivity Resolution0.01, 0.1, 1 μS/cm; 0.01, 0.1 mS/cm
  • Conductivity Accuracy±1.0% full-scale
  • CellK=1.0
  • Cell Constant0.1, 1, 10 cm–1
  • Min TDS0 g/L
  • Max TDS100 μg/L
  • TDS Accuracy±1.0% full scale
  • Min Salinity0 ppt
  • Max Salinity100 ppt
  • Salinity Accuracy±1.0% full scale
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • Min Temperature (° F)32
  • Max Temperature (° F)212
  • Temperature Resolution0.1°F/°C
  • Temperature Accuracy±0.9°F (0.5°C) ±1 digit
  • Temperature compensation0 to 100˚C (manual or automatic)
  • Temperature Coefficient2.0%/°C
  • WaterproofIP57
  • Battery3 AA (included)
  • Battery life (hr)400
  • Length (cm)9.1
  • Width (cm)19
  • Height (cm)3.3
  • Width (in)7 31/64
  • Length (in)3 37/64
  • Height (in)1 19/64
  • Stirring ControlNo
  • NIST Calibration IncludedNo
  • Probe IncludedpH, EC/ATC, and temperature
  • Accessories IncludedNone
  • DescriptionOakton PC100 Portable pH/Conductivity Meter with pH, EC, and Temperature Probes
  • Warranty2 year
Reviews
Q&A