Cole-Parmer Carboy Spigot, fits 06035-81 to 06035-87
Representative image only.
Cole-Parmer Carboy Spigot, fits 06035-81 to 06035-87
Cole-Parmer | Item: UZ-06035-76
Leakproof spigot designed for durable applications
$21.07CAD / Each
BUY MORE & SAVE
  • 1-5$21.07 CAD / Each
  • 6-29$18.96 CAD / Each
  • 30-119$17.91 CAD / Each
  • 120+$16.86 CAD / Each
$21.07CAD / Each
BUY MORE & SAVE
  • 1-5$21.07 CAD / Each
  • 6-29$18.96 CAD / Each
  • 30-119$17.91 CAD / Each
  • 120+$16.86 CAD / Each
Product Description
More About this Item
This replacement spigot is compatible with Cole-Parmer Essentials heavy-walled narrow-mouth HDPE carboys, models 06035-81, -83, -85, and -87. The threaded end of the spigot is 3/4" NPT and the OD of the spigot end is 11.5 mm.
Specifications
  • Bottle ColorTranslucent
  • AutoclavableNo
  • DescriptionCarboy Spigot, fits 06035-81 to 06035-87