The product you searched is discontinued. Below is the suggested replacement. Compare to discontinued item.

Corning Transwell® Permeable Supports, Sterile, Polycarbonate Membrane, 0.4 µm, 24-Cluster Plate; 48/Cs

Representative image only.

Corning Transwell® Permeable Supports, Sterile, Polycarbonate Membrane, 0.4 µm, 24-Cluster Plate; 48/Cs

Corning | Mfr: 3413| Item: 1948007

Essential tool for the study of both anchorage-dependent and independent cell lines

$676.29CAD / Case of 48

$676.29CAD / Case of 48
Product Description

Key Features

  • Provide an environment for cells that closely resembles their in vivo state
  • Treated for optimal cell attachment
  • Ideal for a variety of cell culture applications—chemotaxis, drug transport, tissue remodeling, and invasion studies
  • Supports come sterile and preloaded in cluster plates
  • Sterilized by gamma radiation
  • Packaged 12 inserts in a 24 well plate, 4 plates per case, for a total of 48 inserts per case

More About this Item

These permeable supports, also known as cell culture inserts, are designed to satisfy your diverse cell culture requirements. A suspended design prevents media wicking between the insert and the plate well. It also allows for co-culturing without damaging cells attached to the plate's growth surface. Choose either polycarbonate (PC) or Polyethylene Terephthalate (PET) membrane material. Polycarbonate membranes are translucent while PET membranes are clear, allowing for excellent visibility under phase contrast microscopy.

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

Specifications
  • Cluster Plate Size Number24
  • Well ShapeFlat
  • Culture Area (cm²)0.33
  • Surface TreatmentTC-Treated
  • Lid (Included)Yes
  • SterileYes
  • Membrane MaterialPolycarbonate
  • Membrane Diameter (mm)6.5
  • Height (mm)22.63
  • Length (mm)127.76
  • Width (mm)85.47
  • ColorTranslucent
  • Pore Size (µm)0.4
  • DescriptionPermeable Supports, Sterile, Polycarbonate Membrane, 0.4 µm, 24-Cluster Plate; 48/Cs
Reviews
Q&A