A&D Weighing AD-1690-01 Pipette Leak Tester Replacement Tube Set; 3 Tubes and 1 Adapter
Representative image only.
A&D Weighing AD-1690-01 Pipette Leak Tester Replacement Tube Set; 3 Tubes and 1 Adapter
A&D Weighing | Mfr: AD-1690-01| Item: UZ-07911-51
Replace tubes to ensure accurate leak testing
$309.41CAD / Each
$309.41CAD / Each
Product Description
Includes
Three tubes and one adapter.
Key Features
  • 6 month warranty
More About this Item
Covers three different tip sizes of pipettes, with capacities ranging from 2 to 10,000 µL.
6 Month Warranty
Specifications
  • Compatible WithPipette leak tester 07911-50
  • DescriptionPipette Leak Tester Replacement Tube Set; 3 tubes and 1 adapter