Cole-Parmer® PCR-300 Alpha Cyclers
Easily create, modify and access programs, saving time
  • Flexible—your choice of one, two, or four block capacities in 96-well, 384-well, or mixed formats
  • Independently control each block for simplified temperature and space optimization
  • Software allows quick access to commonly used protocols
These compact thermal cyclers deliver reproducible results every run—with little to no training needed. Features include a clear responsive touch screen, secure user-specific programming, adjustable heated lid, and active sample cooling for sharper amplification.
A Program Wizard generates a protocol specific to your sequence in seconds—a quick way to optimize new assays. Units also retain approximately 1000 reports for reviewing at a later date and run on an intuitive, HD Android™ tablet interface. Save your personal settings to your USB drive; sign in is as simple as inserting your USB drive, no password needed.
$8,545.66 - $34,919.34CAD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Block Description Number of Blocks
Availability Pricing
COMPARE
One 384-well plate 1
$8,545.66CAD / Each
COMPARE
One set of 96 x 0.2 mL tubes or one 96-well plate 1
$8,803.51CAD / Each
COMPARE
Three 384-well plates, one set of 96 x 0.2 mL tubes or one 96-well plate 4
$34,064.78CAD / Each
COMPARE
Two 384-well plates, two sets of 96 x 0.2 mL tubes or two 96-well plates 4
$34,919.34CAD / Each
COMPARE
Four 384-well plates 4
$34,919.34CAD / Each
COMPARE
One 384-well plate, three sets of 96 x 0.2 mL tubes or three 96-well plates 4
$34,919.34CAD / Each
COMPARE
Four sets of 96 x 0.2 mL tubes or four 96-well plates 4
$34,919.34CAD / Each
COMPARE
One 384-well plate, one set of 96 x 0.2 mL tubes or one 96-well plate 2
$19,831.83CAD / Each
COMPARE
Two sets of 96 x 0.2 mL tubes or two 96-well plates 2
$20,207.54CAD / Each
COMPARE
Two 384-well plates 2
$20,207.54CAD / Each
Showing 1- 10 of 10
Need Help with this product?
Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.
Call Us At800-363-5900
Live chat