The Flu? Not You! Ways to Protect Yourself
Antiseptic Hand Cleanser
Antiseptic Hand Cleanser
According to the Centers for Disease Control and Prevention (CDC), flu activity is picking up. In most years, flu activity peaks in January or February but can vary from year to year.  With the winter months approaching, you can take precautions to protect yourself from the virus.
Hand sanitizers can kill bacteria and viruses within 15 seconds of contact. Keep our 64-oz Antiseptic Hand Cleanser nearby, even when soap and water aren’t available. It meets OSHA standards and contains aloe vera to prevent drying. Another option is the Bacdown® Gel No-Rinse Skin Sanitizer. This nondrying antiseptic skin cleanser includes gentle emollients to smooth and moisten skin. The Bacdown Gel reduces bacteria count on hands prior to gloving or entering aseptic environments.
Disposable respirators can provide a barrier from fluid penetration and the transfer of microorganisms. Our ONE-Fit™ Healthcare N95 Particulate Respirators provide 99.9% Bacterial Filtration Efficiency (BFE) and offer 99.5% Particle Filtration Efficiency (PFE), and 99.9% Viral Filtration Efficiency (VFE). They are designed for healthcare settings, such as emergency rooms and intensive care units and meet CDC guidelines for M Tuberculosis.
In addition to using safety products such as these, cover your coughs and sneezes with tissues, keep work surfaces (including telephones and computers) clean, and avoid using coworkers’ phones and computers to help prevent the spread of viruses.
Be the first to comment on "The Flu? Not You! Ways to Protect Yourself"