PCRmax ECO48 Real-time qPCR System: World’s Most Accurate Block – Watch VIDEO
The PCRmax Eco 48 real-time qPCR System utilizes patented block and optical technologies to deliver extreme speed, sensitivity and data quality, with optimized protocol as fast as 15 minutes, all using standard plasticware and consumables, sensitivity down to one copy detection and the world’s most accurate block.
The PCRmax Eco 48 is a high specification, economically priced real-time thermal cycler that accommodates a unique 48-well polypropylene PCR plate utilizing the same geometry as standard 384-well plates, but only 1/8 of the size. This enables users to dramatically reduce the qPCR reagent volumes compared to traditional 96-well instruments, whilst still producing a strong fluorescence signal. Minimizing the plate size also significantly improves thermal uniformity. A minimum volume of 5µl is validated, cutting reagent costs in half and making more efficient use of expensive and hard to acquire template DNA samples.
Watch this short video to find out more.
The Eco 48 Thermal Cycler workflow is based on three simple steps:
  • Load samples into the 48-well plate using the backlit Eco Dock and place the plate into the Eco 48.
  • Run experiment.
  • The Eco 48 software provides instrument control, data collection, and data analysis. The most common qPCR parameters are automatically in place. These can be easily changed on the computer to meet specific experimental requirements.
qPCR System Features and Benefits:
  • MIQE compliant open license software giving confidence when it comes to publishing your data.
  • HRM functionality as standard: with the ability to discriminate class IV SNP >99% of the time.
  • Eco 48 allows for simple multiplex analysis with up to four dyes at once.
  • World’s most thermally accurate block gives consistent data every time.
  • High uniformity provides high-quality data. Fast cycling enables several experiments per day, all at an economical price.
  • Fastest block-based real-time qPCR system. 40 cycles in 40 minutes standardly and 40 cycles in as little as 15 minutes after optimization of temperatures and hold times.
  • Eco 48 is an open platform that can utilize any chemistry, dye or PCR reagent.
  • Easy to use software, making analysis simple and fast.
  • Industry-leading data quality.
  • Eco 48 format provides a small 34.5cm x 31cm (13.6” × 12.2”) footprint to save bench space
Want even more information on PCRmax Eco48 Thermal Cycler. Call our product support specialists at 1-800-323-4340.
View all available thermal cyclers from Cole-Parmer
Be the first to comment on "PCRmax ECO48 Real-time qPCR System: World’s Most Accurate Block – Watch VIDEO"