Silicone Tubing Sizing
Examples of Silicone Tubing Sizing
available at
Dimensions inch (mm)
Internal Diameter
Outside Diameter
Wall thickness
1/50 (0.5)
1/12 (2.1)
1/32 (0.8)
1/32 (0.8)
3/32 (2.4)
1/32 (0.8)
1/16 (1.6)
1/8 (3.2)
1/32 (0.8)
3/32 (2.4)
5/32 (4.0)
1/32 (0.8)
1/8 (3.2)
3/16 (4.8)
1/32 (0.8)
1/8 (3.2)
1/4 (6.4)
1/16 (1.6)
1/8 (3.2)
3/8 (9.6)
1/8 (3.2)
5/32 (4.0)
7/32 (4.0)
1/32 (0.8)
3/16 (4.8)
1/4 (6.4)
1/32 (0.8)
3/16 (4.8)
5/16 (8.0)
1/16 (1.6)
3/16 (4.8)
7/16 (11.2)
1/8 (3.2)
1/4 (6.4)
5/16 (8.0)
1/32 (0.8)
1/4 (6.4)
3/8 (9.6)
1/16 (1.6)
1/4 (6.4)
7/16 (11.2)
3/32 (2.4)
1/4 (6.4)
1/2 (12.8)
1/8 (3.2)
5/16 (8.0)
7/16 (11.2)
1/16 (1.6)
5/16 (8.0)
1/2 (12.8)
3/32 (2.4)
5/16 (8.0)
9/16 (14.4)
1/8 (3.2)