Ponndorf Hose Chemical Compatibility Chart
resistant 1
partially resistant 2
not resistant 3
not tested
Natural rubber = NR
Neoprene = N
Hypalon = H
Perbunan = NBR
= EDPM
= Ponnprene
A | B | C | D | E | F | G | H | I | J | K | L | M | N | O | P | Q | R | S | T | U | V | W | X | Y | Z
Medium NR N H NBR EPDM Ponnnprene
acetaldehyde 3 2 1 3 1 1
acetates
acetic acid 10 % 1 1 2 1 1
acetic acid 25 % 2 2 2 2 1
acetic acid 40 % 3 3 1 3
acetic acid 75 % 3 3 3 3 1
acetic acid, conc. 1 1 2 2 1
acetic acid, vapour 1 1 2 1
acetic anhydride 1 1 1 3 3 3
acetone 1 1 2 3 2 3
acetylene 1 1 1 1 2 1
acrylate dispersion 1 1 1
air (jp to 135°C) 1
alcohol 1 1 1 1 1
aliphatic acid 3 1 2 1 3
aluminum chloride 1 1 1 1 2 1
aluminum fluoride 1 1 1 1 2
aluminum sulfate 1 1 1 1 2
alum 1 1 1 2 1
amine 3 3 2 2
ammonia anhydrous 1 1 1 3 2 1
ammonia, watery 1 1 1 3 1 1
ammonium carbonate 1 1 1 2 1
ammonium chloride 1 1 1 2 1 1
ammonium hydroxide 2 1 1 3 2 1
ammonium nitrate 1 1 1 2 2
ammonium sulfate 1 1 1 1 2
amocool soluble 3 % 1 1
amyl acetate 2 2 3 3 3 3
amyl alcohol 1 1 1 3 2 1
Andol 4,5 H1 36 1
aniline 3 3 3 3 1
aniline-dyes 2 1 2
animal oils 1
antimonic salts
aqua regia 3 3 1 3
arsenic 1 1 1 1
barium chloride 1 1 1 1 2 1
barium hydroxide 1 1 1 1 1 1
barium salts 1 1 1 1 2 1
barium sulfide 1 1 1 1 2
battery acid 2 1 1 2 1
becroson KSM 1
beer 1 1 1 2 2 1
benzaldehyde 3 3 3 3 1
benzene 3 3 3 3 3 3
benzyl alcohol 1 1 1 3 2 1
benzyl chloride 3 3 3 3
benzoic acid 3 3
bitumen 2 1 2 1
blast furnace gas 2 1 1 1
borax 1 1 1 1 1 1
boric acid 1 1 1 2 2 1
brake fluid 1
brega raw oil
go BACK TO TOP