This product is now discontinued, View Related Products
Zetex Aramid and Leather Palm Gloves
Reinforced palms provide excellent abrasion and cut protection
Reinforced gloves offer heat protection up to 800°F (427°C). Reinforced palms are sewn-in and double-stitched for maximum durability. Select aramid palm gloves for additional cut protection. Leather palm gloves feature superior abrasion resistance. One size fits all. Made in the USA.
Approval: UL listed
Compliance: European EN 407 industrial standard
$194.49CAD / Pair of 1
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Material Size
Availability Pricing
Newtex Industries Style 130 Gloves with Leather Palm, 1 pr RK-86364-32 Mfr # 2100009
COMPARE
Leather One size fits all
$194.49CAD / Pair of 1