Zefon Bio-Pump® Plus Pump and TSI® 4046 Calibrator
Fast and easy air samplings with Bio-Pump® Plus pump and Air-O-Cell®/Via-Cell® cassettes
  • Bio-Pump Plus pump is portable and lightweight
  • Ensure sampling pump accuracy with flow calibrator
  • Flow calibrator comes with NIST-traceable certificate
Collect air samples anywhere you need with Bio-Pump Plus pump. This small, battery-powered IAQ pump is designed to be used exclusively with Air-O-Cell and Via-Cell air sampling cassettes for fast sample collection. Pump shows real-time display of actual run time and features "sample complete" audible and visual indicators. Programmable sample time allows unattended operation. Order the Bio-Pump Plus pro kit with additional accessories for complete air sampling.
Easily calibrate your personal sampling pumps before and after each use with the battery-operated TSI® 4046 calibrator. The calibrator has a range of 2.5 to 300 L/min and is accurate to ±2% of reading. Continuous display of volumetric flow rate makes it easy to adjust pump flow in real time. Flow calibration kit includes the TSI 4046 calibrator and all the connections needed to use with Bio-Pump Plus pump. For your convenience, calibration kit 60062-01 includes a Bio-Pump Plus pump, a TSI 4046 calibrator, and accessories—everything you need to ensure flow rate accuracy.
$1,373.20 - $4,001.73CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Calibration Kit with TSI-4046 Calibrator
$3,388.80CAD / Each
COMPARE
Calibration Kit with TSI-4046 Calibrator and Bio-Pump® Plus Pump
$4,001.73CAD / Each
COMPARE
Bio-Pump® Plus IAQ Pump Pro Kit with Accessories
$1,373.20CAD / Each