This product is now discontinued, View Related Products
YSI Model 5100 Dissolved Oxygen Benchtop Meters
Keypad with tactile keys provide audible confirmations to reduce errors
  • Large LCD is easy to read — even from 10 feet away
  • Menu-driven operation makes setup simple
These benchtop meters are easy to use. All meters feature autocalibration, data storage of up to 100 sets, automatic stabilization, and real-time clock. Meters operate on AC power or batteries. Order probes and accessories separately.
The YSI® 5100 calculates the oxygen and specific oxygen uptake rates (OUR and SOUR) via built-in software. Membrane integrity feature indicates when membrane requires replacing. This meter helps you easily comply with U.S. EPA regulations. The built-in barometer automatically compensates readings with no need to enter or correlate barometric pressure values for truly one-button autocalibration.
$2,799.44 - $3,094.12CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Dissolved Oxygen Min Dissolved Oxygen
Availability Pricing
YSI 5100-115 Model 5100 Benchtop Dissolved Oxygen Meter, 115 VAC RK-05510-22 Mfr # 051004
COMPARE
0.0 to 60.0 ppm
$2,799.44CAD / Each
YSI 051005 Model 5100 Benchtop Dissolved Oxygen Meter, 230 VAC RK-05510-27 Mfr # 051005
COMPARE
0.0 to 60.0 ppm
$3,094.12CAD / Each