This product is now discontinued, View Related Products
YSI EcoSense pH100A pH/mV/Temp Meter
Accurate, economical portable meter
  • Automatic buffer recognition for US and NIST buffers
  • Automatic or manual temperature compensation
The pH100A combines low cost with a rugged, waterproof design--making it ideal for spot sampling in wastewater, surface water, or aquaculture. Meter features easy-to-use interface, one-hand operation, automatic or manual temperature compensation, electrode offset recognition, and electrode efficieny display. Meter also has low battery indicator, auto shutoff, and nonvolatile 50 data sets memory with calibration data. Calibrate the meter quickly using the internal buffer recognition feature.
Purchase meter with probes separately to customize your system, or choose a kit for a complete system.
$434.80 - $640.92CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Calibration Max pH (pH) Description
Availability Pricing
YSI PH100 Ecosense pH100A pH/mV/Temp Handheld Meter Only RK-59351-32 Mfr # 606075
COMPARE
Up to 2 points 16 Ecosense pH100A pH/mV/Temp Handheld Meter Only
$434.80CAD / Each
YSI PH100 Ecosense pH100A pH/mV/Temp Handheld Meter Kit RK-59351-34 Mfr # 606067
COMPARE
Up to 2 points 16 Ecosense pH100A pH/mV/Temp Handheld Meter Kit
$640.92CAD / Each