This product is now discontinued, View Related Products
YSI Ecosense EC300A Conductivity Meter
Accurate, economical portable meter with four-electrode conductivity cell
  • Rugged, weighted probe assembly with automatic temperature compensation
  • Measures conductivity, salinity or TDS with temperature
The EC300A features easy-to-use interface, one-hand operation, and waterproof housing with rugged, weighted probes that can be ordered with 13- or 33-ft (4- or 10-m) cable. The rugged, four-electrode conductivity cell provides stable measurements from 0 to 200 mS with auto-ranging function that eliminates the need for multiple conductivity cell constants. Meter also offers auto shutoff, low battery indicator, and nonvolatile 50 data sets memory with calibration data.
Purchase meter with probes separately to customize your system, or choose a kit for a complete system.
$500.95 - $1,082.94CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Conductivity Resolution Min Conductivity Description
Availability Pricing
YSI EC300 Ecosense Conductivity Meter, without Probes RK-19750-30 Mfr # 606079
COMPARE
0.1 μS or 0.1 mS 0.0 to 200 mS Ecosense Conductivity Meter, without Probes
$500.95CAD / Each
YSI EC300 Ecosense A Conductivity Meter Kit RK-19750-32 Mfr # 601036
COMPARE
0.1 μS or 0.1 mS 0.0 to 200 mS Ecosense A Conductivity Meter Kit
$1,082.94CAD / Each