This product is now discontinued, View Related Products
WTW ProfiLine Oxi 3210 Handheld Dissolved Oxygen Meters
A rugged line of DO meters with a range of features, benefits, and price points to meet every application and budget!
  • Datalogging for up to 200 manual readings and expanded salinity compensation range
  • Galvanic probes require no warm-up time
  • Rugged, waterproof housing is great for the field or the lab
Durable, I67-rated meters feature a 100% waterproof and easy-to-clean keypad. Silicone rubber keys provide tactile feedback for error-free operation. The large graphic display allows for excellent readability even under difficult lighting conditions. Convenient meter kits provide all the accessories needed to start taking readings. Choose kits with CellOx® 325 probes for general applications such as surface water monitoring or laboratory measurements. DurOx® 325 probes offer additional durability and are for applications such as wastewater aeration monitoring.
Meters only include: CD-ROM and four AA batteries.
Meter kits add: specified probe, cleaning solution, electrolyte, replacement membrane heads, stand, beaker, and hard carrying case.
$1,265.64 - $2,431.09CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Min Dissolved Oxygen Min Salinity Temperature compensation
Availability Pricing
WTW Oxi 3210 ProfiLine Dissolved Oxygen Meter Only RK-53105-35 Mfr # 2BA200
COMPARE
0 to 90.0 ppm 0 to 70 ppt Automatic
$1,265.64CAD / Each
WTW Oxi 3210 ProfiLine Dissolved Oxygen Meter w/CellOx 325 Probe, 1.5 m RK-53105-36 Mfr # 2BA201
COMPARE
0 to 90.0 ppm 0 to 70 ppt Automatic
$2,431.09CAD / Each
WTW Oxi 3210 ProfiLine Dissolved Oxygen Meter w/CellOx 325 Probe, 3 m RK-53105-37 Mfr # 2BA202
COMPARE
0 to 90.0 ppm 0 to 70 ppt Automatic
$2,224.82CAD / Each
WTW Oxi 3210 ProfiLine Dissolved Oxygen Meter w/DurOx Probe, 3 m RK-53105-38 Mfr # 2BA203
COMPARE
0 to 90.0 ppm 0 to 70 ppt Automatic
$2,033.28CAD / Each