This product is now discontinued, View Related Products

WTW ProfiLine Oxi 3205 Handheld Dissolved Oxygen Meters

A rugged line of DO meters with a range of features, benefits, and price points to meet every application and budget!
  • Designed for users who desire a simple-to-use, yet dependable instrument
  • Galvanic probes require no warm-up time
  • Rugged, waterproof housing is great for the field or the lab
Durable, I67-rated meters feature a 100% waterproof and easy-to-clean keypad. Silicone rubber keys provide tactile feedback for error-free operation. The large graphic display allows for excellent readability even under difficult lighting conditions. Convenient meter kits provide all the accessories needed to start taking readings. Choose kits with CellOx® 325 probes for general applications such as surface water monitoring or laboratory measurements. DurOx® 325 probes offer additional durability and are for applications such as wastewater aeration monitoring.

Meters only include: CD-ROM and four AA batteries.

Meter kits add: specified probe, cleaning solution, electrolyte, replacement membrane heads, stand, beaker, and hard carrying case.
$1,162.50 - $2,180.62CAD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Min Dissolved Oxygen Min Salinity Probe Included
Availability Pricing
WTW Oxi 3205 ProfiLine Dissolved Oxygen Meter Only RK-53105-30 Mfr # 2BA100
COMPARE
0 to 90.0 ppm 0 to 35 ppt No
$1,162.50CAD / Each
WTW 2BA101 ProfiLine Oxi 3205 Dissolved Oxygen Meter w/CellOx 325 Probe, 1.5 m RK-53105-31 Mfr # 2BA101
COMPARE
0 to 90.0 ppm 0 to 35 ppt Yes
$2,180.62CAD / Each
WTW Oxi 3205 ProfiLine Dissolved Oxygen Meter w/CellOx 325 Probe, 3 m RK-53105-33 Mfr # 2BA102
COMPARE
0 to 90.0 ppm 0 to 35 ppt
$2,124.63CAD / Each
WTW Oxi 3205 ProfiLine Dissolved Oxygen Meter w/DurOx Probe, 3 m RK-53105-34 Mfr # 2BA103
COMPARE
0 to 90.0 ppm 0 to 35 ppt
$1,673.77CAD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat