This product is now discontinued, View Related Products
WTW OxiTop IS BOD Self-Check Measurement Systems
Eliminate daily checks and increase data reliability with automated BOD testing
  • Data is automatically stored in the measuring head
  • Readings are given directly in mg/L
  • Sample dilution is not required, saving you time
  • AutoTemp function delays start of test until sample temperature stabilizes
These OxiTop automated BOD measurement systems provide a ready-to-use solution for both small and large laboratories. Choose the OxiTop IS system when a small number of tests need to be performed. Systems are available in 6- and 12-place configurations and include everything you need to begin testing.
Convenient and simple to use — place the OxiTop measuring head directly on the BOD bottle containing the sample. The head senses pressure changes caused by cellular respiration within the closed system and converts that data into BOD values. The measurements are then stored in the imbedded memory for later retrieval.
The included IS stirrer base maintains constant temperature and distributes oxygen evenly throughout the sample. Order the optional thermostat box to incubate samples at 20°C; the box measures 16-3/4"W x 14-3/4"H x 23-5/8"D (42.5 x 37.5 x 60 cm) and can accommodate an entire 12-place OxiTop system.
OxiTop IS System uses the OxiTop measuring head with an LED display. The head automatically stores one value every 24 hours for five days. This system is best suited for fewer tests since programming and data recording must be done by hand.
$4,405.44 - $7,175.41CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
WTW OxiTop IS BOD Measurement System, 6 Samples RK-59354-02 Mfr # 208210
COMPARE
BOD Measurement System, 6 Samples
$4,405.44CAD / Each
WTW OxiTop IS BOD Measurement System, 12 Samples RK-59354-04 Mfr # 208211
COMPARE
BOD Measurement System, 12 Samples
$7,175.41CAD / Each
WTW 208212 BOD OXITOP IS 6 TO 12 RK-59354-06 Mfr # 208212
COMPARE
$5,038.99CAD / Each