Cole-Parmer® Essentials Write-On Colored Tape
Identify specific applications by color coding
  • Withstands temperatures from –10 to 250 ºF (–23 to 121 ºC)
  • Waterproof, and oil and acid resistant
Use tape to identify specific applications by color coding. This durable labeling tape is waterproof as well as oil and acid resistant. Tape can be used with pencil, pen, or permanent marker.
$59.67 - $107.56CAD / Pkg of 6
24 variations of this product are available.
See All Product Options Below
24 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Length (ft) Width (in)
Availability Pricing
COMPARE
White 42 1/2
$80.30CAD / Pkg of 6
COMPARE
Yellow 42 1/2
$86.19CAD / Pkg of 6
COMPARE
Green 42 1/2
$84.72CAD / Pkg of 6
COMPARE
Pink 42 1/2
$86.19CAD / Pkg of 6
COMPARE
Red 42 1/2
$84.72CAD / Pkg of 6
COMPARE
Orange 42 1/2
$86.19CAD / Pkg of 6
COMPARE
Blue 42 1/2
$80.30CAD / Pkg of 6
COMPARE
Assorted 42 1/2
$107.56CAD / Pkg of 24
COMPARE
White 42 3/4
$64.09CAD / Pkg of 4
COMPARE
Yellow 42 3/4
$78.09CAD / Pkg of 4
COMPARE
Green 42 3/4
$76.62CAD / Pkg of 4
COMPARE
Pink 42 3/4
$76.62CAD / Pkg of 4
COMPARE
Red 42 3/4
$73.67CAD / Pkg of 4
COMPARE
Orange 42 3/4
$78.09CAD / Pkg of 4
COMPARE
Blue 42 3/4
$78.09CAD / Pkg of 4
COMPARE
Assorted 42 3/4
$95.03CAD / Pkg of 16
COMPARE
White 42 1
$76.62CAD / Pkg of 3
COMPARE
Yellow 42 1
$76.62CAD / Pkg of 3
COMPARE
Green 42 1
$59.67CAD / Pkg of 3
COMPARE
Pink 42 1
$66.30CAD / Pkg of 3
Showing 1 - 20 of 24