Worthington (aka Taylor-Wharton) Cryogenic Shippers
Safely transport your cryogenic samples!
  • Shippers provide long liquid nitrogen holding times
  • Adsorbent filler prevents spills and leaks
Sturdy aluminum shell and positive-closure cap protect contents. Inner storage canister is constructed of heavy-duty stainless steel..
$2,741.98 - $7,954.83CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Volume (Liters)
Availability Pricing
Worthington CX100B-11M Cryogenic Shippers, 3.7 L, 30 day static hold time RK-44300-08 Mfr # CX100B-11M
COMPARE
3.7
$2,741.98CAD / Each
Worthington CXR500-0 Cryogenic Shippers, 6.4 L, 14 day static hold time RK-44300-10 Mfr # CXR500-0
COMPARE
6.4
$7,954.83CAD / Each